Learn More
Cayman Chemical FCCP, 370-86-5, 10 mg
SDP
Supplier:  Cayman Chemical 1521810
Encompass_Preferred
A potent uncoupler of oxidative phosphorylation in mitochondria that disrupts ATP synthesis by transporting protons across cell membranes
Catalog No. NC0904863
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.