Learn More
Medchemexpress LLC HY-100410 100mg , FCCP Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone CAS:370-86-5 Purity:98%
SDP
Catalog No. 501493261
Encompass_Preferred
Medchemexpress, HY-100410 100mg FCCP Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone CAS:370-86-5 Formula:C10H5F3N4O Purity:98% FCCP is a mitochondrial uncoupling agent, used in cancer research. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3261 Supplier Medchemexpress LLC Supplier No. HY100410100MG
Medchemexpress, HY-100410 100mg FCCP Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone CAS:370-86-5 Formula:C10H5F3N4O Purity:98% FCCP is a mitochondrial uncoupling agent, used in cancer research. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.