Learn More
Enzo Life Sciences FCCP (50mg). CAS: 370-86-5
SDP
Supplier:  Enzo Life Sciences BMLCM1200050
Encompass_Preferred
A potent reversible inhibitor of mitochondrial oxidative phosphorylation. FCCP is a useful tool for depolarizing mitochondrial membrane potential. Treatment of cells with FCCP at varying concentrations leads to partial (100 nM) or complete (10 µM) depolarization and apoptosis. Alternative name: Carbonylcyanide-4-(trifluoromethoxy)-phenylhydrazone. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: Ambient.
Catalog No. 50-201-0426