Learn More
Abcam FCCP, mitochondrial o x idative phosphorylation uncoupler, 50MG
Catalog No. NC0907735
Encompass_Preferred
FCCP uncoupler, mitochondrial oxidative phosphorylation is with purity of > 99%. - Cited in over 40 publications - Soluble in DMSO to 100 mM
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0907735 Supplier Abcam Supplier No. AB12008150MG
FCCP uncoupler, mitochondrial oxidative phosphorylation is with purity of > 99%. - Cited in over 40 publications - Soluble in DMSO to 100 mM
The product is subject to the following: Abcam Restricted Use Statement