Learn More
Abcam FCCP, mitochondrial o x idative phosphorylation uncoupler, 10MG
Supplier: Abcam AB12008110MG
FCCP uncoupler, mitochondrial oxidative phosphorylation is with purity of > 99%. - Cited in over 40 publications - Soluble in DMSO to 100 mM
The product is subject to the following: Abcam Restricted Use Statement