Learn More
ACROBiosystems ACROBiosystems Human Fc epsilon RI alpha Protein, Fc Tag (BLI verified)
Catalog No. 501956431
Encompass_Preferred
Recombinant Protein;100UG;Human Fc epsilon RI alpha, Fc Tag (BLI verified) (FCA-H5259) is expressed from human 293 cells (HEK293). It contains AA Val 26 - Leu 204 (Accession # P12319-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 47.3 kDa. The protein migrates as 55-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6431 Supplier ACROBiosystems Supplier No. FCAH5259100UG
Recombinant Protein;100UG;Human Fc epsilon RI alpha, Fc Tag (BLI verified) (FCA-H5259) is expressed from human 293 cells (HEK293). It contains AA Val 26 - Leu 204 (Accession # P12319-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 47.3 kDa. The protein migrates as 55-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.