Learn More
Aviva Systems Biology Corp FCER1A Antibody - N-terminal region (ARP45203_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45203_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FCER1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FCER1A. Alternate names: FCE1A, FcERIApplicationWB. Species reactivity: Human
Catalog No. 50-014-63060