Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus Fc gamma RI / CD64 Protein, His,Avitag™ (BLI verified)
Supplier:  ACROBiosystems FCAC82E825UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Cynomolgus CD64, His,Avitag (BLI verified) (FCA-C82E8) is expressed from human 293 cells (HEK293). It contains AA Val 11 - Pro 288 (Accession # NP_001270969.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 34.8 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized FMC63 at 1 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus CD64, His,Avitag (BLI verified) (Cat. No. FCA-C82E8) with a linear range of 0.2-10 ng/mL (Routinely tested).
Catalog No. 50-195-6882