Learn More
ACROBiosystems ACROBiosystems Rhesus macaque Fc gamma RIIA / CD32a Protein, His Tag
Catalog No. 501956391
Encompass_Preferred
Recombinant Protein;1MG;Rhesus macaque CD32a, His Tag (CDA-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 34 - Ile 217 (Accession # NP_001244229.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.3 kDa.
Catalog No. 50-195-6391 Supplier ACROBiosystems Supplier No. CDAR52H41MG
Recombinant Protein;1MG;Rhesus macaque CD32a, His Tag (CDA-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 34 - Ile 217 (Accession # NP_001244229.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.3 kDa.