Learn More
ACROBiosystems ACROBiosystems Cynomolgus Fc gamma RIIA / CD32a Protein, His Tag (SPR & BLI & HPLC verified)
Catalog No. 501956582
Encompass_Preferred
Recombinant Protein;50UG;Cynomolgus CD32a, His Tag (SPR & BLI & HPLC verified) (CDA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Pro 208 (Accession # Q8SPW4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.1 kDa. The protein migrates as 27-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Captured Cynomolgus CD32a, His Tag (SPR & BLI & HPLC verified) (Cat. No. CDA-C52H7) on CM5 Chip via Anti-His antibody can bind Rituximab with an affinity constant of 4.18 μM as determined in SPR assay (Biacore T200) (QC tested).
Catalog No. 50-195-6582 Supplier ACROBiosystems Supplier No. CDAC52H750UG
Recombinant Protein;50UG;Cynomolgus CD32a, His Tag (SPR & BLI & HPLC verified) (CDA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Pro 208 (Accession # Q8SPW4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.1 kDa. The protein migrates as 27-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Captured Cynomolgus CD32a, His Tag (SPR & BLI & HPLC verified) (Cat. No. CDA-C52H7) on CM5 Chip via Anti-His antibody can bind Rituximab with an affinity constant of 4.18 μM as determined in SPR assay (Biacore T200) (QC tested).