Learn More
ACROBiosystems ACROBiosystems Human Fc gamma RIIA / CD32a (R167) Protein, His Tag (BLI verified)
Supplier:  ACROBiosystems CDAH522150UG
Encompass_Preferred
Recombinant Protein;50UG;Human CD32a (R167), His Tag (BLI verified) (CDA-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 36 - Ile 218 (Accession # P12318-1 (R167)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.6 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD32a (R167), His Tag (BLI verified) (Cat. No. CDA-H5221) on CM5 Chip via anti-His antibody, can bind  Rituximab with an affinity constant of 3.12 μM as determined in a SPR assay (Biacore T200) (Routinely tested).
Catalog No. 50-195-7460