Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus Fc gamma RIIA / CD32a Protein, His,Avitag™ (SPR & BLI verified)
Catalog No. 501956886
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Cynomolgus CD32a, His,Avitag (SPR & BLI verified) (CDA-C82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Pro 208 (Accession # Q8SPW4-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 23.9 kDa. The protein migrates as 30-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Captured Biotinylated Cynomolgus CD32a, His,Avitag (SPR & BLI verified) (Cat. No. CDA-C82E5) on Biotin CAP - Series S sensor Chip can bind MabThera (Rituximab) with an affinity constant of 7.55 μM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. 50-195-6886 Supplier ACROBiosystems Supplier No. CDAC82E5200UG
Recombinant Protein;200UG;Biotinylated Cynomolgus CD32a, His,Avitag (SPR & BLI verified) (CDA-C82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Pro 208 (Accession # Q8SPW4-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 23.9 kDa. The protein migrates as 30-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Captured Biotinylated Cynomolgus CD32a, His,Avitag (SPR & BLI verified) (Cat. No. CDA-C82E5) on Biotin CAP - Series S sensor Chip can bind MabThera (Rituximab) with an affinity constant of 7.55 μM as determined in a SPR assay (Biacore T200) (QC tested).