Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Fc gamma RIIA / CD32a (H167), His Tag, Avi Tag (Avitag™) (SPR verified);HEK293;200UG;CDA,CD32a,FCGR2A,CD32,FCG2 ,FCGR2A1,IGFR2,Fcr,Fcgr,Fc receptor,Fc gamma receptor
Catalog No. 501659172
Encompass_Preferred
Recombinant Protein;Biotinylated Human Fc gamma RIIA / CD32a (H167), His Tag, Avi Tag (Avitag™) (SPR verified);200UG;Biotinylated Human CD32a (H167), His Tag, Avi Tag (SPR verified) (CDA-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ala 36 - Ile 218 (Accession # P12318-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 24.0 kDa. The protein migrates as 30-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Biotinylated Human CD32a (H167), His Tag, Avi Tag (Cat. No. CDA-H82E6) captured on Biotin CAP- Series S Sensor Chip can bind Rituximab with an affinity constant of 0.66 μM as determined in a SPR assay (Biacore T200) (QC tested).;Immobilized Biotinylated Human CD32a (H167), His Tag, Avi Tag (Cat. No. CDA-H82E6) on SA Chip can bind Rituximab with an affinity constant of 0.74 μM as determined in a SPR assay (Biacore T200) (Routinely tested).
Catalog No. 50-165-9172 Supplier ACROBiosystems Supplier No. CDAH82E6200UG
Recombinant Protein;Biotinylated Human Fc gamma RIIA / CD32a (H167), His Tag, Avi Tag (Avitag™) (SPR verified);200UG;Biotinylated Human CD32a (H167), His Tag, Avi Tag (SPR verified) (CDA-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ala 36 - Ile 218 (Accession # P12318-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 24.0 kDa. The protein migrates as 30-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Biotinylated Human CD32a (H167), His Tag, Avi Tag (Cat. No. CDA-H82E6) captured on Biotin CAP- Series S Sensor Chip can bind Rituximab with an affinity constant of 0.66 μM as determined in a SPR assay (Biacore T200) (QC tested).;Immobilized Biotinylated Human CD32a (H167), His Tag, Avi Tag (Cat. No. CDA-H82E6) on SA Chip can bind Rituximab with an affinity constant of 0.74 μM as determined in a SPR assay (Biacore T200) (Routinely tested).