Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus Fc gamma RIIB / CD32b Protein, His,Avitag™ (SPR & BLI verified)
Catalog No. 501956889
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Cynomolgus CD32b, His,Avitag (SPR & BLI verified) (CDB-C82E4) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 224 (Accession # Q8SPW3-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 23.6 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Captured Biotinylated Cynomolgus CD32b, His,Avitag (SPR & BLI verified) (Cat. No. CDB-C82E4) on Biotin CAP - Series S sensor Chip can bind MabThera (Rituximab) with an affinity constant of 4.94 μM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. 50-195-6889 Supplier ACROBiosystems Supplier No. CDBC82E425UG
Recombinant Protein;25UG;Biotinylated Cynomolgus CD32b, His,Avitag (SPR & BLI verified) (CDB-C82E4) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 224 (Accession # Q8SPW3-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 23.6 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Captured Biotinylated Cynomolgus CD32b, His,Avitag (SPR & BLI verified) (Cat. No. CDB-C82E4) on Biotin CAP - Series S sensor Chip can bind MabThera (Rituximab) with an affinity constant of 4.94 μM as determined in a SPR assay (Biacore T200) (QC tested).