Learn More
ACROBiosystems Recombinant Protein;Mouse Fc gamma RIIB / CD32b Protein (SPR verified);HEK293;100UG;CDB,FCGR2B,CD32b,CD32,FCG2,IGFR2,CDw32,FCG-2,IGFR-2,Fcr,Fcgr,Fc receptor,Fc gamma receptor
Supplier:  ACROBiosystems CDBM52H7100UG
Encompass_Preferred
Recombinant Protein;Mouse Fc gamma RIIB / CD32b Protein (SPR verified);100UG;Mouse CD32b (SPR verified) (CDB-M52H7) is expressed from human 293 cells (HEK293). It contains AA Thr 40 - Arg 217 (Accession # NP_001070657).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.2 kDa. The protein migrates as 32-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse CD32b (Cat. No. CDB-M52H7) on NTA-Series S sensor chip , can bind OKT3 with an affinity constant of 0.86 μM as determined in a SPR assay (Biacore T200) (QC tested).;Immobilized OKT3 on Protein G-Series S sensor chip, can bind Mouse CD32b (Cat. No. CDB-M52H7) with an affinity constant of 0.55 μM as determined in a SPR assay (Biacore T200) (Routinely tested).
Catalog No. 50-165-7520