Learn More
ACROBiosystems Recombinant Protein;Biotinylated Mouse Fc gamma RIIB / CD32b Protein, His Tag, Avi Tag (Avitag™) (SPR verified);HEK293;200UG;CDB,FCGR2B,CD32b,CD32,FCG2,IGFR2,CDw32,FCG-2,IGFR-2,Fcr,Fcgr,Fc receptor,Fc gamma receptor
Supplier:  ACROBiosystems CDBM82E8200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Mouse Fc gamma RIIB / CD32b Protein, His Tag, Avi Tag (Avitag™) (SPR verified);200UG;Biotinylated Mouse CD32b, His Tag, Avi Tag (SPR verified) (CDB-M82E8) is expressed from human 293 cells (HEK293). It contains AA Thr 40 - Arg 217 (Accession # NP_001070657.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 24.0 kDa. The protein migrates as 38-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Biotinylated Mouse CD32b, His Tag, Avi Tag (SPR verified) (Cat. No. CDB-M82E8) captured on Biotin CAP- Series S Sensor Chip can bind OKT3 with an affinity constant of 1.53 μM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. 50-165-7452