Learn More
ACROBiosystems ACROBiosystems Cynomolgus Fc gamma RIII / CD16 Protein, His Tag (BLI verified)
Supplier:  ACROBiosystems FC6C52H950UG
Encompass_Preferred
Recombinant Protein;50UG;Cynomolgus CD16, His Tag (BLI verified) (FC6-C52H9) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # Q8SPW2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.8 kDa. The protein migrates as 37-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rituximab at 5 μg/mL (100 μL/well) can bind Cynomolgus CD16, His Tag (BLI verified) (Cat. No. FC6-C52H9) with a linear range of 39-313 ng/mL (QC tested).;Captured Cynomolgus CD16, His Tag (BLI verified) (Cat. No. FC6-C52H9) on CM5 chip via anti-His antibody can bind MabThera (Rituximab) with an affinity constant of 0.206 μM as determined in a SPR assay (Biacore T200) (Routinely tested).
Catalog No. 50-195-7238