Learn More
ACROBiosystems ACROBiosystems Mouse Fc gamma RIII / CD16 Protein, His Tag (SPR & BLI verified)
Supplier:  ACROBiosystems CDAM52H850UG
Encompass_Preferred
Recombinant Protein;50UG;Mouse CD16, His Tag (SPR & BLI verified) (CDA-M52H8) is expressed from human 293 cells (HEK293). It contains AA Leu 32 - Thr 215 (Accession # Q5D5I8).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.0 kDa. The protein migrates as 36-45 kDa under reducing (R) condition (SDS-PAGE).;Mouse CD16, His Tag (SPR & BLI verified) (Cat. No. CDA-M52H8) Captured on NTA-Series S sensor chip can bind OKT3 with an affinity constant of 0.126 μM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. 50-195-7269