Learn More
ACROBiosystems ACROBiosystems Rhesus macaque Fc gamma RIII / CD16 Protein, His Tag
Supplier:  ACROBiosystems FC6R52H650UG
Encompass_Preferred
Recombinant Protein;50UG;Rhesus macaque CD16, His Tag (FC6-R52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Gly 206 (Accession # A3RFZ7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.1 kDa.
Catalog No. 50-195-6390