Learn More
ACROBiosystems ACROBiosystems Human Fc gamma RIIIA / CD16a (V176) Protein, His Tag (SPR & BLI verified)
Supplier:  ACROBiosystems CD8H52H450UG
Encompass_Preferred
Recombinant Protein;50UG;Human CD16a (V176), His Tag (SPR & BLI verified) (CD8-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # P08637-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.7 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD16a (V176), His Tag (SPR & BLI verified) (Cat. No. CD8-H52H4) on CM5 Chip via anti-His antibody, can bind  Herceptin with an affinity constant of 163 nM as determined in SPR assay (Biacore T200) (QC tested).
Catalog No. 50-195-7252