Learn More
ACROBiosystems Recombinant Protein;Human Fc gamma RIIIA / CD16a (F176) Protein (SPR verified);HEK293;1MG;CDA,FCGR3A,CD16A,FCG3,FCGR3,IGFR3,FcRIIIa,FcRIII,Fc-gamma RIII,Fc-gamma RIIIa,Fcr,Fcgr,Fc receptor,Fc gamma receptor
Catalog No. 501657807
Encompass_Preferred
Recombinant Protein;Human Fc gamma RIIIA / CD16a (F176) Protein (SPR verified);1MG;Human CD16a (F176) (SPR verified) (CDA-H5220) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # P08637-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.7 kDa. The protein migrates as 39-51 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD16a (F176) (Cat. No. CDA-H5220) on CM5 Chip via anti-His antibody, can bind  Rituximab with an affinity constant of 0.63 μM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. 50-165-7807 Supplier ACROBiosystems Supplier No. CDAH52201MG
Recombinant Protein;Human Fc gamma RIIIA / CD16a (F176) Protein (SPR verified);1MG;Human CD16a (F176) (SPR verified) (CDA-H5220) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # P08637-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.7 kDa. The protein migrates as 39-51 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD16a (F176) (Cat. No. CDA-H5220) on CM5 Chip via anti-His antibody, can bind  Rituximab with an affinity constant of 0.63 μM as determined in a SPR assay (Biacore T200) (QC tested).