Learn More
ACROBiosystems ACROBiosystems Human Fc gamma RIIIB / CD16b (NA2) Protein, His Tag (SPR & BLI verified)
Catalog No. 501957458
Encompass_Preferred
Recombinant Protein;50UG;Human CD16b (NA2), His Tag (SPR & BLI verified) (CDB-H5222) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Ser 200 (Accession # O75015-1).The NA1 form of the CD16b differ with the NA2 form of CD16b in AA36, 65, 82, and 106. The NA1 form carries R36, N65, D82, and V106, while the NA2 form carries S36, S65, N82, and I106.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.7 kDa. The protein migrates as 38-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD16b (NA2), His Tag (SPR & BLI verified) (Cat. No. CDB-H5222) on CM5 Chip via anti-His antibody, can bind Rituximab with an affinity constant of 2.88 μM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. 50-195-7458 Supplier ACROBiosystems Supplier No. CDBH522250UG
Recombinant Protein;50UG;Human CD16b (NA2), His Tag (SPR & BLI verified) (CDB-H5222) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Ser 200 (Accession # O75015-1).The NA1 form of the CD16b differ with the NA2 form of CD16b in AA36, 65, 82, and 106. The NA1 form carries R36, N65, D82, and V106, while the NA2 form carries S36, S65, N82, and I106.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.7 kDa. The protein migrates as 38-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD16b (NA2), His Tag (SPR & BLI verified) (Cat. No. CDB-H5222) on CM5 Chip via anti-His antibody, can bind Rituximab with an affinity constant of 2.88 μM as determined in a SPR assay (Biacore T200) (QC tested).