Learn More
ACROBiosystems Recombinant Protein;Human Fc gamma RIIIB / CD16b (NA1) Protein (SPR verified);HEK293;100UG;CDB,Fc gamma RIIIB / CD16b (NA1),FCGR3B,CD16B,FCG3B,FCGR3,FCG3,IGFR3,FCGR-3,FCG-3,IGFR-3,Fcr,Fcgr,Fc receptor,Fc gamma receptor
Catalog No. 501658361
Encompass_Preferred
Recombinant Protein;Human Fc gamma RIIIB / CD16b (NA1) Protein (SPR verified);100UG;Human CD16b (NA1) (SPR verified) (CDB-H5227) is expressed from human HEK293 cells. It contains AA Gly 17 - Ser 200 (Accession # AAA35881.1).The NA1 form of the CD16b differ with the NA2 form of CD16b in AA36, 65, 82, and 106. The NA1 form carries R36, N65, D82, and V106, while the NA2 form carries S36, S65, N82, and I106.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.8 kDa. The protein migrates as 35-45 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD16b (NA1) (Cat. No. CDB-H5227) on CM5 Chip via anti-His antibody , can bind Rituximab with an affinity constant of 8.17 μM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. 50-165-8361 Supplier ACROBiosystems Supplier No. CDBH5227100UG
Recombinant Protein;Human Fc gamma RIIIB / CD16b (NA1) Protein (SPR verified);100UG;Human CD16b (NA1) (SPR verified) (CDB-H5227) is expressed from human HEK293 cells. It contains AA Gly 17 - Ser 200 (Accession # AAA35881.1).The NA1 form of the CD16b differ with the NA2 form of CD16b in AA36, 65, 82, and 106. The NA1 form carries R36, N65, D82, and V106, while the NA2 form carries S36, S65, N82, and I106.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.8 kDa. The protein migrates as 35-45 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD16b (NA1) (Cat. No. CDB-H5227) on CM5 Chip via anti-His antibody , can bind Rituximab with an affinity constant of 8.17 μM as determined in a SPR assay (Biacore T200) (QC tested).