Learn More
ACROBiosystems Recombinant Protein;Mouse Fc gamma RIII / CD16 Protein;HEK293;100UG;CDA,FCGR3,Fc-gamma RIII,IgG Fc receptor III,Fc gamma Receptor III,Fc gamma RIII,FcgRIII
Catalog No. 501658726
Encompass_Preferred
Recombinant Protein;Mouse Fc gamma RIII / CD16 Protein;100UG;Mouse CD16 (CDA-M52H8) is expressed from human 293 cells (HEK293). It contains AA Leu 32 - Thr 215 (Accession # Q5D5I8).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.95 kDa. The protein migrates as 36-45 kDa under reducing (R) condition (SDS-PAGE).;Immobilized OKT3 at 10 μg/mL (100 μL/well) can bind Mouse CD16 (Cat. No. CDA-M52H8) with a linear range of 78-625 ng/mL (QC tested).
Catalog No. 50-165-8726 Supplier ACROBiosystems Supplier No. CDAM52H8100UG
Recombinant Protein;Mouse Fc gamma RIII / CD16 Protein;100UG;Mouse CD16 (CDA-M52H8) is expressed from human 293 cells (HEK293). It contains AA Leu 32 - Thr 215 (Accession # Q5D5I8).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.95 kDa. The protein migrates as 36-45 kDa under reducing (R) condition (SDS-PAGE).;Immobilized OKT3 at 10 μg/mL (100 μL/well) can bind Mouse CD16 (Cat. No. CDA-M52H8) with a linear range of 78-625 ng/mL (QC tested).