Learn More
ACROBiosystems Recombinant Protein;Cynomolgus Fc gamma RIII / CD16 Protein;HEK293;100UG;FC6,FCGR3,Fc-gamma RIII,IgG Fc receptor III,Fc gamma Receptor III,Fc gamma RIII,FcgRIII
Catalog No. 501658123
Encompass_Preferred
Recombinant Protein;Cynomolgus Fc gamma RIII / CD16 Protein;100UG;Cynomolgus CD16 (FC6-C52H9) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # Q8SPW2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.9 kDa. The protein migrates as 37-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rituximab at 5 μg/mL (100 μL/well) can bind Cynomolgus CD16 (Cat. No. FC6-C52H9) with a linear range of 39-313 ng/mL (QC tested).
Catalog No. 50-165-8123 Supplier ACROBiosystems Supplier No. FC6C52H9100UG
Recombinant Protein;Cynomolgus Fc gamma RIII / CD16 Protein;100UG;Cynomolgus CD16 (FC6-C52H9) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # Q8SPW2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.9 kDa. The protein migrates as 37-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rituximab at 5 μg/mL (100 μL/well) can bind Cynomolgus CD16 (Cat. No. FC6-C52H9) with a linear range of 39-313 ng/mL (QC tested).