Learn More
ACROBiosystems Recombinant Protein;Biotinylated Cynomolgus Fc gamma RIII / CD16 Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;FC6,FCGR3,Fc-gamma RIII,IgG Fc receptor III,Fc gamma Receptor III,Fc gamma RIII,FcgRIII
Catalog No. 501658121
Encompass_Preferred
Recombinant Protein;Biotinylated Cynomolgus Fc gamma RIII / CD16 Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Cynomolgus CD16, His Tag, Avi Tag (FC6-C82E0) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # Q8SPW2-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 25.7 kDa. The protein migrates as 36-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rituximab at 5 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus CD16, His Tag, Avi Tag (Cat. No. FC6-C82E0) with a linear range of 0.039-1.25 μg/mL (QC tested).
Catalog No. 50-165-8121 Supplier ACROBiosystems Supplier No. FC6C82E025UG
Recombinant Protein;Biotinylated Cynomolgus Fc gamma RIII / CD16 Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Cynomolgus CD16, His Tag, Avi Tag (FC6-C82E0) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # Q8SPW2-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 25.7 kDa. The protein migrates as 36-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rituximab at 5 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus CD16, His Tag, Avi Tag (Cat. No. FC6-C82E0) with a linear range of 0.039-1.25 μg/mL (QC tested).