Learn More
ACROBiosystems ACROBiosystems Mouse Fc gamma RIV / CD16-2 Protein, His Tag
Supplier:  ACROBiosystems FC4M52H31MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse CD16-2, His Tag (FC4-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Gln 203 (Accession # A0A0B4J1G0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.8 kDa. The protein migrates as 28-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Captured Mouse CD16-2, His Tag (Cat. No. FC4-M52H3) on NTA-Series S sensor chip can bind OKT3 with an affinity constant of 154 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
Catalog No. 50-195-6951