Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse Fc gamma RIV / CD16-2 Protein, His,Avitag™ (BLI verified)
Supplier:  ACROBiosystems FC4M82E8200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Mouse Fc gamma RIV, His,Avitag (BLI verified) (FC4-M82E8) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Gln 203 (Accession # A0A0B4J1G0-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 24.6 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized FMC63 at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse Fc gamma RIV, His,Avitag (BLI verified) (Cat. No. FC4-M82E8) with a linear range of 0.3-20 ng/mL (Routinely tested).
Catalog No. 50-195-6804