Learn More
Aviva Systems Biology Corp FCGR2A Antibody - C-terminal region (ARP63808_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63808_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FCGR2A. It was validated on Western Blot. . Alternate names: CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A1, FcGR, IGFR2, MGC23887, MGC30032ApplicationWB. Species reactivity: Human
Catalog No. 50-288-091