Learn More
Aviva Systems Biology Corp FCGR2B Antibody - C-terminal region (ARP63532_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63532_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FCGR2B. It was validated on Western Blot. . Alternate names: CD32, CD32B, FCG2, FCGR2, IGFR2ApplicationWB. Species reactivity: Human
Catalog No. 50-288-000