Learn More
Aviva Systems Biology Corp FCGR3A Antibody - middle region (ARP63559_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63559_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FCGR3A. It was validated on Western Blot. . Alternate names: CD16A, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-011