Learn More
ABclonal Technology FCGR3B Rabbit pAb
SDP
Supplier:  ABclonal Technology A139801000UL
Encompass_Preferred
The protein encoded by this gene is a low affinity receptor for the Fc region of gamma immunoglobulins (IgG). The encoded protein acts as a monomer and can bind either monomeric or aggregated IgG. This gene may function to capture immune complexes in the peripheral circulation. Several transcript variants encoding different isoforms have been found for this gene. A highly-similar gene encoding a related protein is also found on chromosome 1.
Catalog No. 50-001-06313