Learn More
Enzo Life Sciences CD16/CD32 Recombinant monoclonal antibody (2.4G2) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4100200
Encompass_Preferred
Clone: 2.4G2. Isotype: IgGκ. Immunogen: Mouse BALB/c Macrophage J774 Cell Line. Species reactivity: Mouse. Specificity: Recognizes an epitope on the extracellular domains of the mouse FcγIII and FcγII, thereby inhibiting their ability to bind antibodies via their Fc region. Also binds the FcγI receptor (CD64) via its Fc domain. Applications: Flow Cytometry, IF, IHC, IP, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2937