Learn More
Aviva Systems Biology Corp FCGRT Antibody - N-terminal region (ARP46406_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46406_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FCGRT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FCGRT. Alternate names: FCRN, alpha-chainApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Sheep
Catalog No. 50-014-53476