Learn More
Sino Biological Recombinant Human FCGRT & B2M Heterodimer Protein (His Tag), HPLC-verified
Supplier:  Sino Biological CT009H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Ser297) of human FCGRT (P55899-1) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the human B2M (P61769) (Met1-Met119) constructed the plasmid 2. The two plasmids were co-expressed and the FCGRT / B2M heterodimer was purified.
Catalog No. 50-211-6323