Learn More
Sino Biological Mouse FCGRT & B2M Heterodimer Protein 1mg
Supplier:  Sino Biological CT029M08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ser 297) of mouse FCGRT (Q61559) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the mouse B2M (P01887,105Ala/Asp) (Met 1-Met 119) constructed the plasmid 2. The two plasmids were co-expressed and the FCGRT / B2M heterodimer was purified
Catalog No. 50-211-6397