Learn More
Sino Biological Cynomolgus FCGRT & B2M Heterodimer Protein (His & AVI Tag), Biotinylated 100ug
Supplier:  Sino Biological CT084C27HB100
Encompass_Preferred
A DNA sequence encoding the cynomolgus FCGRT (Q8SPV9) (Met 1-Ser 297) was fused with a c-terminal polyhistidine tagged AVI tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the cynomolgus B2M (Q8SPW0) (Met 1-Met 119) constructed the plasmid 2. The two plasmids were co-expressed and the FCGRT/B2M heterodimer was purified. The purified heterodimer was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5381