Learn More
ACROBiosystems FCGRT&B2M HETERODIMER PROTEIN
Supplier:  ACROBiosystems FCMC52W950UG
Encompass_Preferred
FCM-C52W9-50ugCynomolgus/Rhesus macaque FcRn/FCGRT&B2M Heterodimer Protein, His-TaggedFormulationLyophilized from 0.22 um filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
Catalog No. NC1751088