Learn More
Aviva Systems Biology Corp FCHO1 Antibody - middle region (ARP55174_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55174_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FCHO1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FCHO1. Alternate names: KIAA0290ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53749