Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCLA Free Acid [Chemiluminescence Reagent], TCI America™
SDP

Catalog No. A531010MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A531010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A531010MG Supplier TCI America Supplier No. A531010MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 133118-06-6
Molecular Formula C36H27N5O7S
MDL Number MFCD04037231
Synonym 3,7-Dihydro-6-[4-[2-[N′C-(5-fluoresceinyl)thioureido]ethoxy]phenyl]-2-methylimidazo[1,2-a]pyrazin-3-one

Specifications

Quantity 10 mg
Formula Weight 673.70
Physical Form Crystalline Powder
Chemical Name or Material FCLA Free Acid [Chemiluminescence Reagent]
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.