Learn More
Aviva Systems Biology Corp FCN1 Antibody - middle region (ARP54605_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54605_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FCN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FCN1. Alternate names: FCNMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-61256