Learn More
Sino Biological FCN1 / FCNA Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10930MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FCN1 / FCNA (rh FCN1 / FCNA; Catalog#10930-H08H; NP_001994.2; Met1-Ala326). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8440