Learn More
Med Vet International Chemistry Reagent Deionized Water Reagent Grade / ASTM Type I 100% 20 Liter, 15335-20L, Each
Catalog No. 502224007
Encompass_Preferred
Strict quality controls ensure this high purity reagent grade water meets or exceeds ASTM Type I, CAP/CLSI clinical laboratory reagent water and USP-NF purified water criteria This high purity reagent grade water is free of all organic and inorganic contaminants and is packaged at 18 megaohms-cm resistivity, the absolute limit for completely pure water There are 20 liters of deionized water in this Cubitainer
Catalog No. 50-222-4007 Supplier Med Vet International Supplier No. 940736EA
Strict quality controls ensure this high purity reagent grade water meets or exceeds ASTM Type I, CAP/CLSI clinical laboratory reagent water and USP-NF purified water criteria This high purity reagent grade water is free of all organic and inorganic contaminants and is packaged at 18 megaohms-cm resistivity, the absolute limit for completely pure water There are 20 liters of deionized water in this Cubitainer