Learn More
Medchemexpress LLC FCPR03 | 1917347-65-9 | 99.8% | 299.31 g/mol | C15H19F2NO3 | 50 MG
SDP
Catalog No. 5000226643
Encompass_Preferred
FCPR03 is a selective phosphodiesterase 4 (PDE4) inhibitor for research use. The compound has CAS 1917347-65-9, formula C15H19F2NO3, molecular weight 299.31 g/mol, and is reported at 99.82% purity. Supplied as a 50 mg quantity, it is intended for in vitro laboratory studies.
  • Selective PDE4 inhibitor with low-nanomolar potency.
  • Reported purity 99.82% suitable for research applications.
  • Supplied as a 50 MG quantity for laboratory use.
  • Soluble in DMSO and compatible with common in vitro assay formats.
  • Analytical data such as COA and solubility guidance available.
Catalog No. 50-002-26643 Supplier Medchemexpress LLC Supplier No. HY11797750MG
FCPR03 is a selective phosphodiesterase 4 (PDE4) inhibitor for research use. The compound has CAS 1917347-65-9, formula C15H19F2NO3, molecular weight 299.31 g/mol, and is reported at 99.82% purity. Supplied as a 50 mg quantity, it is intended for in vitro laboratory studies.
  • Selective PDE4 inhibitor with low-nanomolar potency.
  • Reported purity 99.82% suitable for research applications.
  • Supplied as a 50 MG quantity for laboratory use.
  • Soluble in DMSO and compatible with common in vitro assay formats.
  • Analytical data such as COA and solubility guidance available.