Learn More
Med Vet International 5-5 1/2" Economy Thumb Tissue Forceps, 1x2 Teeth
Catalog No. 502223969
Encompass_Preferred
Thumb forceps are spring forceps used by compression between your thumb and forefinger and are used for grasping, holding or manipulating body tissue. They are non-ratchet style. For example, you could use thumb forceps to hold or move tissue during surgery or to move dressings.
Catalog No. 50-222-3969 Supplier Med Vet International Supplier No. 8920
Thumb forceps are spring forceps used by compression between your thumb and forefinger and are used for grasping, holding or manipulating body tissue. They are non-ratchet style. For example, you could use thumb forceps to hold or move tissue during surgery or to move dressings.