Learn More
Cayman Chemical FcRIIIb/CD16b HNA-1b allotyp
SDP
Catalog No. 502947306
Encompass_Preferred
Source: Recombinant human FcγRIIIb expressed in E. coli • Amino acids: 18-193 • MW: 20 kDa
Catalog No. 50-294-7306 Supplier Cayman Chemical Supplier No. 31827100 UG
Source: Recombinant human FcγRIIIb expressed in E. coli • Amino acids: 18-193 • MW: 20 kDa