Learn More
Aviva Systems Biology Corp FCRL1 Antibody - N-terminal region (ARP50228_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50228_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FCRL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FCRL1. Alternate names: DKFZp667O1421, FCRH1, IFGP1, IRTA5, RP11-367J7.7, CD307aApplicationWB. Species reactivity: Cow, Horse, Human, Mouse
Catalog No. 50-014-60293