Learn More
Aviva Systems Biology Corp FCRLB Antibody - N-terminal region (ARP62552_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62552_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FCRLB. It was validated on Western Blot. . Alternate names: FCRL2, FCRLM2, FCRLY, FLJ31052, FREB-2, FcRY, RP11-474I16.6ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-711