Learn More
BPS Bioscience Inc FCRN IgG Recycling HMEC-1 Cell Pool, Cell Pool, HMEC-1, 2 vials, >1 x 10^6 cells per vial, 1 ml of 7.5% DMSO and 92.5% FBS, Research Use Only
SDP
Catalog No. NC2868568
Encompass
FCRN IgG Recycling HMEC-1 Cell Pool, Cell Pool, HMEC-1, 2 vials, >1 x 10^6 cells per vial, 1 ml of 7.5% DMSO and 92.5% FBS, Research Use Only
Catalog No. NC2868568 Supplier BPS Bioscience Inc Supplier No. 82163
Add to Cart
Add to Cart
Request a Quote
FCRN IgG Recycling HMEC-1 Cell Pool, Cell Pool, HMEC-1, 2 vials, >1 x 10^6 cells per vial, 1 ml of 7.5% DMSO and 92.5% FBS, Research Use Only