Learn More
ACROBiosystems Biotinylated Canine FcRn (FCGRT & B2M) Heterodimer Protein, His & Avitag, 200 μg (FCM-C82W9)
Catalog No. 502357327
Encompass_Preferred
Canine FcRn (FCGRT & B2M) heterodimer protein, enzymatically biotinylated and produced in HEK293 cells, supplied as a 200 μg lyophilized vial. Purity and molecular weight are verified by SDS-PAGE and SEC-MALS, and the product is formulated for stability in lyophilized form for use in binding and biophysical assays.
  • Enzymatically biotinylated for site-specific labeling.
  • Co-expressed heterodimer of FCGRT and B2M in HEK293 cells.
  • Supplied as a 200 μg lyophilized vial for long-term stability.
  • Purity greater than 95% confirmed by SDS-PAGE and SEC-MALS.
  • Suitable for SPR, SEC-MALS, and ELISA applications.
  • Includes defined amino acid ranges for each subunit.
Catalog No. 50-235-7327 Supplier ACROBiosystems Supplier No. FCMC82W9200UG
Canine FcRn (FCGRT & B2M) heterodimer protein, enzymatically biotinylated and produced in HEK293 cells, supplied as a 200 μg lyophilized vial. Purity and molecular weight are verified by SDS-PAGE and SEC-MALS, and the product is formulated for stability in lyophilized form for use in binding and biophysical assays.
  • Enzymatically biotinylated for site-specific labeling.
  • Co-expressed heterodimer of FCGRT and B2M in HEK293 cells.
  • Supplied as a 200 μg lyophilized vial for long-term stability.
  • Purity greater than 95% confirmed by SDS-PAGE and SEC-MALS.
  • Suitable for SPR, SEC-MALS, and ELISA applications.
  • Includes defined amino acid ranges for each subunit.